TISSUE SPECIFICITY: Predominantly expressed in trigeminal and dorsal root sensory ganglia. Isoform 1 and isoform 3 are also expressed in brain and peripheral blood mononuclear cells.
Immunohistochemical analysis of rat DRG indicates a superb degree of specificity for this serum.
Application Information:
IHC. Use at 1:1000 to 1:2000 dilution. This antibody has not been tested in other applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: capsaicin receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human capsaicin receptor protein (aa: 531-541)
Host: Rabbit
Isotype: IgG
Reactivity: Rat