Anti-mouse PERK Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-1333-100EA 312 EUR
BSENR-1333-100
Anti-mouse PERK Rabbit Polyclonal Antibody
Vasta-aineet
PERK (PKR-like ER kinase) is a single-pass type I ER membrane protein with a stress-sensing luminal domain connected by a transmembrane segment to a cytoplasmic-kinase domain.

Antibody is known to recognize mouse PERK both phosphorylated and non-phosphorylated forms by Western Blotting (WB) and Immunoprecipitation (IP). A dilution of 1:500 is recommended for WB. A dilution of 30 µl of antibody in a total reaction mixture of 500 µl is recommended for IP. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
Western Blotting (WB) and Immunoprecipitation (IP). A dilution of 1:500 is recommended for WB. A dilution of 30 µl of antibody in a total reaction mixture of 500 µl is recommended for IP. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: mouse PERK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR