Coronins belong to the WD40 or WD family of proteins. Coronins appear to be particularly involved in binding to actin, actin associated proteins, tubulin and phospholipase C and have been implicated in the mechanisms of chemotaxis and phagocytosis. In mammals there are at least five major coronin proteins, named coronins 1 to 5 in one nomenclature. Another nomenclature divides these five proteins in coronins 1a and 1b, 2a, 2b and 2c (see the Human Genone Organization Gene Nomenclature Committee link for this family). The mammalian coronin family members are abundant components of eukaryotic cells and each type has a restricted cell type specific expression pattern. Coronin 1A is found exclusively in hematopoetic lineage cells such as lymphocytes, macrophages and neutrophils. This antibody is therefore an excellent marker of cells of this lineage and can also be used to study the leading edges particularly of neutrophils. Since the only hematopoetic cells found within the central nervous system are microglia, this antibody is also an excellent marker of this important cell type. Microglia are numerically fairly minor components of the nervous system, but microglial activation is seen in response to a wide variety of damage and disease states, including ALS, Alzheimer's disease and responses to brain tumors. Since coronin 1a is a constitutive component of microglia, the coronin 1a antibody can be used to study both quiescent and activated microglia.
The specificity of this antibody has been confirmed by WB and ICC against the antigen.
Application Information:
Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A concentration of 1:10,000-20,000 is recommended for WB. Human Coronin 1A has a predicted length of 461 residues and a MW of 51 kDa. A concentration of 1:500-1:1,000 is recommended for IC and IH. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Coronin 1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse, Cat