Anti-ATG12 Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-137-500EA 302 EUR
BSENR-137-500
Anti-ATG12 Rabbit Polyclonal Antibody
Vasta-aineet
FUNCTION: Required for autophagy. SUBUNIT: Conjugated to ATG5. SUBCELLULAR LOCATION: Cytoplasm. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Ubiquitous. SIMILARITY: Belongs to the ATG12 family.

IHC and WB confirmed the specificity for ATG12.
Application Information:
IHC, immunofluorescence, WB. A working concentration of 1-5 µg/ml is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal of Human ATG12
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR