Anti-GABARAP L2 Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-142-100EA 334 EUR
BSENR-142-100
Anti-GABARAP L2 Rabbit Polyclonal Antibody
Vasta-aineet
FUNCTION: Involved in intra-Golgi traffic. Modulates intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF which in turn stimulates the association with GOSR1. SUBUNIT: Monomer. Interacts with GABRG2, NSF, GOSR1 and beta-tubulin. Interacts with ULK1. SUBCELLULAR LOCATION: Golgi apparatus. TISSUE SPECIFICITY: Ubiquitous. Expressed at high levels in the brain, heart, prostate, ovary, spleen and skeletal muscle. Expressed at very low levels in lung, thymus and small intestine. SIMILARITY: Belongs to the MAP1 LC3 family.

IHC, WB and ELISA confirmed the specificity for GABARAP L2 (GABARAPL2). A 14kDa band, that corresponds to the molecular weight of GABARAPL2, is detected via western blot analysis.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:200 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GABARAP L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal of Human GABARAP L2 (GABARAPL2)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR