Anti-GABARAP Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-143-100EA 302 EUR
BSENR-143-100
Anti-GABARAP Rabbit Polyclonal Antibody
Vasta-aineet
GABARAP is highly positively charged in its N-terminus and shares sequence homology with MAP1LC3 1A and 1B. This protein clusters neurotransmitter receptors (GABA(A) receptors) by mediating interaction with the cytoskeleton.  SUBUNIT: Interacts with GABRG2, TUBA1, ULK1 and NSF. Interacts with beta-tubulin and GPHN. SUBCELLULAR LOCATION: Intracytoplasmic membrane. Cytoskeleton. Largely associated with intracellular membrane structures including the Golgi apparatus and post-synaptic cisternae. Colocalizes with microtubules. TISSUE SPECIFICITY: Heart, brain, placenta, skeletal muscle, kidney and pancreas.

IHC, WB and ELISA confirmed the specificity for GABARAP.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:200 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal of Human GABARAP
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR