Anti-MAP1LC3 C Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-140-100EA 302 EUR
BSENR-140-100
Anti-MAP1LC3 C Rabbit Polyclonal Antibody
Vasta-aineet
FUNCTION: Probably involved in formation of autophagosomal vacuoles (autophagosomes). SUBUNIT: 3 different light chains, LC1, LC2 and LC3, can associate with MAP1A and MAP1B proteins. SUBCELLULAR LOCATION: LC3-I: Cytoplasm. LC3-II: Intracytoplasmic membrane; lipid-anchor. LC3-II binds to the autophagic membranes. TISSUE SPECIFICITY: Most abundant in placenta, lung and ovary. PTM: The precursor molecule is cleaved by APG4B/ATG4B to form the cytosolic form, LC3-I. This is activated by APG7L/ATG7, transferred to ATG3 and conjugated to phospholipid to form the membrane-bound form, LC3-II. SIMILARITY: Belongs to the MAP1 LC3 family.

IHC and WB confirmed the specificity for MAP1LC3 C . This antibody should recognise MAP1LC3 C only and not the other forms MAP1LC3A and MAP1LC3B.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:100 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAP1LC3 C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal of Human MAP1LC3 C
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR