FUNCTION: Functions within a multiprotein E3 ubiquitin ligase complex, catalyzing the covalent attachment of ubiquitin moieties onto substrate proteins. These substrates include SYT11, CCNE1, GPR37, STUB1, a 22 kDa O-linked glycosylated isoform of SNCAIP and SEPT5. May play a more general role in the ubiquitin proteasomal pathway by participating in the removal and/or detoxification of abnormally folded or damaged protein. Loss of this ubiquitin ligase activity appears to be the mechanism underlying pathogenesis of PARK2. May protect neurons against alpha synuclein toxicity, proteasomal dysfunction, GPR37 accumulation, and kainate-induced excitotoxicity. May play a role in controlling neurotransmitter trafficking at the presynaptic terminal and in calcium-dependent exocytosis. Regulates cyclin E during neuronal apoptosis. May represent a tumor suppressor gene. SUBCELLULAR LOCATION: Cytoplasm. Co-localizes with STY11 in neutrites. Co-localizes with SNCAIP in brainstem Lewy bodies. TISSUE SPECIFICITY: Highly expressed in the brain including the substantia nigra. Expressed in heart, testis and skeletal muscle. Expression is down-regulated or absent in tumor biopsies, and absent in the brain of PARK2 patients. Overexpression protects dopamine neurons from kainate-mediated apoptosis.
This antiserum iis known to specifically recognise Parkin shown by IHC and WB.
Application Information:
IHC, WB, immunoblot, 1-site ELISA. A dilution of 1:500 to 1:2000 is recommended for these applications. This antiserum stains trigeminal motor neurons in rat brain stem. A 50 kDa band was identified in rat brain extract using western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Parkin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig, Rat