- Koti
- Vasta-aineet
- Anti-Contactin-6 Rabbit Polyclonal Antibody
Anti-Contactin-6 Rabbit Polyclonal Antibody
Toimittaja: Biosensis
BSENR-127-100EA
302
EUR
BSENR-127-100
Anti-Contactin-6 Rabbit Polyclonal Antibody
Vasta-aineet
The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development. Employing Northern blot analysis Kamei et al found that amongst different regions of the adult human nervous system cerebellum expressed highest level of NB-3 mRNA. The expression of NB-3 in the cerebellum increases until adulthood. In contrast, the expression in the cerebrum declines to a low level after postnatal day 7. NB-3 like other neural recognition molecules plays a vitally important role in axonal guidance during development, plasticity, and maintenance of synaptic connections in the adult brain. Cui et al recently showed that NB-3 acts as a novel Notch ligand to participate in oligodendrocyte generation. Furthermore, NB-3 triggers nuclear translocation of the Notch intracellular domain and promotes oligodendrogliogenesis from progenitor cells and differentiation of oligodendrocyte precursor cells via Deltex1. In primary oligodendrocytes, NB-3 increases myelin-associated glycoprotein transcripts. Hence, the NB-3/Notch signaling pathway may be worthwhile a closer examination for its potential for the treatment of demyelinating diseases. Human NB-3 shares with rat NB-3 86% identity in nucleotide sequences and 90% identity in amino acid sequences. FUNCTION: Contactins mediate cell surface interactions during nervous system development. Participates in oligodendrocytes generation by acting as a ligand of NOTCH1. Its association with NOTCH1 promotes NOTCH1 activation through the released notch intracellular domain (NICD) and subsequent translocation to the nucleus. Involved in motor coordination. SUBCELLULAR LOCATION: Cell membrane; lipid-anchor; GPI-anchor. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Expressed in brain. In brain, it is preferentially expressed in the accessory olfactory bulb, layers II/III and V of the cerebral cortex, piriform cortex, anterior thalamic nuclei, locus coeruleus of the pons and mesencephalic trigeminal nucleus and in Purkinje cells of the cerebellum. DEVELOPMENTAL STAGE: Highly expressed after birth, reaching a maximum at the postnatal day 7, and declines thereafter in the cerebrum, whereas it increases in the cerebellum to adulthood.
This antiserum stains beautifully the NB3 in DRG, where it nicely stains a subpopulation of neurons and in the trigeminal nerve root, where staining is restricted to the oligodendrocytes.
Application Information:
IHC. A dilution of 1:1000 to 1: 5000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
IHC. A dilution of 1:1000 to 1: 5000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Contactin-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Antigen: Contactin-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Tiedustele hintaa asiakaspalvelusta.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Tuote on lisätty koriin