Anti-HA-Tag Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-1307-100EA 436 EUR
BSENR-1307-100
Anti-HA-Tag Rabbit Polyclonal Antibody
Vasta-aineet
The Human influenza hemagglutin (HA) tag corresponds to a region (98-106 amino acids) from the HA molecule.

Antibody detects HA-tagged proteins (on amino- and carboxy termini) in transfected mammalian cells.
Application Information:
Western Blotting at a suggested dilution 1:500~1:1000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: HA-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: HA epitope
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR