Neurturin (NTN) is a member of the GDNF family of neurotrophic factors. This protein is a potent survival factor for several populations of central and peripheral neurons in mature and developing rodents. FUNCTION: Supports the survival of sympathetic neurons in culture. May regulate the development and maintenance of the CNS. Might control the size of non-neuronal cell population such as haemopoietic cells. SUBUNIT: Homodimer; disulfide-linked. SUBCELLULAR LOCATION: Secreted protein. DISEASE: Defects in NRTN are a cause of Hirschsprung disease (HSCR). In association with mutations of RET gene, and possibly with other loci, defects in NRTN are involved in Hirschsprung disease. This genetic disorder of neural crest development is characterized by the absence of intramural ganglion cells in the hindgut, often resulting in intestinal obstruction. SIMILARITY: Belongs to the TGF-beta family. GDNF subfamily.
Dot blot shows no cross reactivity to GDNF.
Application Information:
IHC, WB, immunoblot, 1-site ELISA. Recommended to be used at a dilution of 1:2000-3000 for immunohistochemistry and Western blot, 1: 2000 to 1:4000 for ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: rh NTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat