Anti-VRL-1 Rabbit Polyclonal Antibody
Toimittaja: Biosensis
BSENR-079-100EA 334 EUR
BSENR-079-100
Anti-VRL-1 Rabbit Polyclonal Antibody
Vasta-aineet
TISSUE SPECIFICITY: Ubiquitously expressed. Expressed in dorsal root ganglia, trigeminal ganglia, spinal chord (Lissauer's tract, dorsal horn and dorsal columns) (at protein level). PTM: N-glycosylated. PTM: Phosphorylated by PKA. SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. SIMILARITY: Contains 3 ANK repeats.
Immunohistochemical analysis in rat dorsal root ganglia and spinal cord indicates a high level of specificity for this antiserum. Specificity was also shown by Western blot.
Application Information:
IHC, Immunofluorescence, Western blot. Recommended to be used at a dilution of 1: 1000 to 1: 2000 for immunohistochemistry and Western blot, for Immunofluorescence at a dilution of 1:50 to 1: 200 in free-floating sections or paraffin embedded sections. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: VRL-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: rat VRL-1 protein (aa: 744-761)
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human