Anti-HAP-1 Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-090-100EA 302 EUR
BSENR-090-100
Anti-HAP-1 Rabbit Polyclonal Antibody
Vasta-aineet
FUNCTION: Associates specifically with huntingtin. This binding is enhanced by an expanded polyglutamine repeat. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: In the brain, especially in the olfactory bulb and in the brain stem. No detectable expression in peripheral tissues such as lung, testis, spleen, and small intestine. SIMILARITY: Contains 1 HAP1 N-terminal domain.

Specificity for HAP-1 was confirmed by IHC.
Application Information:
IHC. A dilution of 1:500 to 1:3000 is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HAP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR