Anti-rh CNTF Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-100-100EA 302 EUR
BSENR-100-100
Anti-rh CNTF Rabbit Polyclonal Antibody
Vasta-aineet
CNTF is a survival promoting factor for different types of neurons in vitro and in vivo. The essential structural features for the biological function of human CNTF were investigated by Thier, M. et al. They showed that deletion of 14 N-terminal and 18 C-terminal amino acids significantly increased bioactivity compared to wild-type CNTF. FUNCTION: CNTF is a survival factor for various neuronal cell types. Seems to prevent the degeneration of motor axons after axotomy. SUBUNIT: Homodimer. SUBCELLULAR LOCATION: Cytoplasm. TISSUE SPECIFICITY: Nervous system. PHARMACEUTICAL: CNTF is being tested under the name Axokine by Regeneron Pharmaceuticals for treatment of human motor neuron diseases, such as amyotrophic lateral sclerosis (ALS). As it induces substantial weight loss, preferentially of fat as opposed to lean body mass, it is being used for obesity treatment. SIMILARITY: Belongs to the CNTF family.

This antibody specifically detects CNTF shown by western blot.
Application Information:
WB. A dilution of 1:500 to 1: 4000 is recommended for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: rh CNTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR