Anti-Semaphorin 5A Mouse Polyclonal Antibody

BSENMO-938-50EA 327 EUR
BSENMO-938-50
Anti-Semaphorin 5A Mouse Polyclonal Antibody
Members of the semaphorin protein family, such as Semaphorin 5A, are involved in axonal guidance during neural development. Semaphorin 5A acts as a bifunctional guidance cue, exerting both attractive and inhibitory effects on developing axons of the fasciculus retroflexus (FR), a diencephalon fiber tract associated with limbic function. Neuronal responses to Semaophorin 5A are dependent on the bound proteoglycan and the biologic context. Mammalian plexin B3 is a high-affinity Semaphorin 5A receptor.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR