SorCS2 is a family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. It is noted that all mammalian VPS10 domains carry an N-terminal signal peptide for translocation into the endoplasmic reticulum. The greatest similarity between the receptors is found in the C-terminal parts of the VPS10 domains, which harbor 12 cysteine residues with a characteristic spacing conserved in all VPS10 proteins from yeast to man. SorCS2 is a single-pass type I membrane protein that is highly expressed in brain and kidney. It is also detected at low levels in heart, liver, small intestine, skeletal muscle and thymus.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: SorCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human SorCS2 (951-1050)
Host: Mouse
Isotype:
Reactivity: Human