Anti-Semaphorin 4G Mouse Polyclonal Antibody

Toimittaja: Biosensis
BSENMO-953-50EA 327 EUR
BSENMO-953-50
Anti-Semaphorin 4G Mouse Polyclonal Antibody
Vasta-aineet
Semaphorins are a large family of conserved secreted and membrane associated proteins which possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. There are eight classes of Semaphorins. Semaphorin 4G is a transmembrane protein that may play a role in axon guidance. It belongs to the semaphorin family.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Semaphorin 4G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Semaphorin 4G (24-115)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR