Prokineticin-2 is expressed in the suprachiasmatic nucleus (SCN) circadian clock and may function as the output component of the circadian clock. The secreted form of the encoded protein may also serve as a chemoattractant for neuronal precursor cells in the olfactory bulb. Mutations in this gene are associated with Kallmann syndrome 4. These patients have variable degrees of olfactory and reproductive dysfunction.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Prokineticin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Prokineticin-2, isoform 2(20-109)
Host: Mouse
Isotype:
Reactivity: Human