Anti-Semaphorin 4A Mouse Polyclonal Antibody

Toimittaja: Biosensis
BSENMO-955-50EA 327 EUR
BSENMO-955-50
Anti-Semaphorin 4A Mouse Polyclonal Antibody
Vasta-aineet
Semaphorin 4A is a transmembrane protein that inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. Defects  in Semaphorin 4A cause retinitis pigmentosa which leads to degeneration of retinal photoreceptor cells. Defects in Semaphorin 4A are also the cause of cone-rod dystrophy type 10.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Semaphorin 4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Semaphorin 4A (33-132)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR