Anti-Cyan Fluorescent Protein Mouse Monoclonal Antibody [clone: B1218]

Toimittaja: Biosensis
BSENM-1300-100EA 341 EUR
BSENM-1300-100
Anti-Cyan Fluorescent Protein Mouse Monoclonal Antibody [clone: B1218]
Vasta-aineet
Recognizes native and denatured forms of Cyan Fluorescent Protein (CFP) and its variants: Enhanced Green Fluorescent Protein (EGFP), Yellow Fluorescent Protein (YFP), Enhanced Yellow Fluorescent Protein (EYFP) and Cyan Fluorescent Protein (CFP).

Recognizes native and denatured forms of Cyan Fluorescent Protein (CFP) and its variants: Enhanced Green Fluorescent Protein (EGFP), Yellow Fluorescent Protein (YFP), Enhanced Yellow Fluorescent Protein (EYFP) and Cyan Fluorescent Protein (CFP).
Application Information:
Western Blotting, ELISA, Dot Blot, IPP, Immunohistochemistry. Suggested dilution of 1:1000 for Western Blotting. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Cyan Fluorescent Protein
Clonality: Monoclonal
Clone: B1218
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR