Anti-Peroxiredoxin-5 Rabbit Polyclonal Antibody
Toimittaja: Biosensis
BSENR-1595-100EA 312 EUR
BSENR-1595-100
Anti-Peroxiredoxin-5 Rabbit Polyclonal Antibody
Vasta-aineet
Peroxiredoxin-5 has a role in intracellular redox signaling.
Antibody reacts with Peroxiredoxin-5 in multiple applications including IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 4,000 is recommended. A dilution of 1: 1,000 to 1: 4,000 is recommended for ELISA. This antibody stains non-ciliated bronchiolar cells in rat lung and what appears to be sebaceous glands in rat skin. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Application Information:
IHC, IF, WB. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 2,000 is recommended. This antibody clearly detects a protein at approximately 22 kDa on WB of human brain tissue. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Peroxiredoxin-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: (190-204 aa) of Human Peroxiredoxin-5
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,