Anti-Presenilin 1 Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-1605-500EA 312 EUR
BSENR-1605-500
Anti-Presenilin 1 Rabbit Polyclonal Antibody
Vasta-aineet
Presenilin-1 (PSEN1) is a multi-pass membrane protein and component of the gamma-secretase complex. PSEN1 is thought to play a role in intracellular signaling and gene expression or in linking chromatin to the nuclear membrane. It may also play a role in hematopoiesis. Defects in PSEN1 are a cause of Alzheimer disease type 3 (AD3), a familial early-onset form of Alzheimer disease (Ref:SWISS-Prot).

Specificity confirmed by WB and IF using transfected cells, Presenilin 1 knock-out mouse cells, mouse and human brain.
Application Information:
IF and WB. Suggested dilution of 1:2,000 is recommended for WB. On SDS-PAGE, the predominant form detected by this antibody is the N-terminal Presenilin 1 fragment of approx 29 kDa. The uncleaved form of Presenilin 1 migrates to approx 45 kDa. Human and mouse brain samples commonly prepared with reducing agent (50mM DTT), urea (2.3M), SDS (1%) in 62.5 mM Tris-HCl pH 6.8 sample buffer heated to 50C for 15 min. The suggested dilution for IF is 1:100 for acetone or paraformaldehyde fixed cells or tissue. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Presenilin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminus of Human Presenilin 1
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR