Anti-homeobox Protein DLX-5 Mouse Monoclonal Antibody [clone: 1B3]

BSENM-825-100EA 327 EUR
BSENM-825-100
Anti-homeobox Protein DLX-5 Mouse Monoclonal Antibody [clone: 1B3]
Homeobox protein DLX-5 belongs to the the distal-less homeobox family. It contains one homeobox DNA-binding domain and is thought to play a role in bone development and fracture healing. It is first expressed at 8.5 to 9 days in facial and branchial arch mesenchyme, otic vesicles and frontonasal ectoderm around olfactory placodes. At day 9.5-10, expression is seen in the developing forebrain. In day 12.5 embryos it is expressed in the brain and bones and also in all skeletal structures of mid gestation embryos after the first cartilage formation. Expression remains unaltered in both brain and skeleton in day 15 embryos and slowly decreases in day 17 embryos. The unprocessed precursor has a length of  289 amino acids and a predicted molecular weight of 31.5 kDa.

The specificity of this antibody was confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for direct and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: homeobox Protein DLX-5
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR