Anti-Brn-3C Mouse Monoclonal Antibody [clone: 5B8]

BSENM-859-100EA 327 EUR
BSENM-859-100
Anti-Brn-3C Mouse Monoclonal Antibody [clone: 5B8]
Brn-3C may play a role in determining or maintaining the identities of a small subset of visual system neurons. It is expressed in the brain and it seems to be specific to the retina. Defects in Brn-3C are the cause of non-syndromic sensorineural deafness autosomal dominant type 15 (DFNA15) which is a form of sensorineural hearing loss. Sensorineuraldeafness results from damage to the neural receptors of the inner ear, the nerve pathways to the brain, or the area of the brain that receives sound information.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, immunofluorescence and sandwich ELISA. The recommended dilution for this antibody is 10 µg/ml for IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Brn-3C
Clonality: Monoclonal
Clone: 5B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR