Anti-hHRT3 Mouse Monoclonal Antibody [clone: 4A11]

BSENM-894-100EA 327 EUR
BSENM-894-100
Anti-hHRT3 Mouse Monoclonal Antibody [clone: 4A11]
hHRT3 belongs to HESR family of basic helix-loop-helix type transcription factors. It contains a bHLH and orange domain, but its YPRW motif has diverged from other HESR family members. It is believed to be a downstream effector of Notch signaling which may be required for cardiovascular development.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, immunofluorescence and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: hHRT3
Clonality: Monoclonal
Clone: 4A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR