Anti-RAPGEFL1 Mouse Monoclonal Antibody [clone: 4G2]

BSENM-897-100EA 327 EUR
BSENM-897-100
Anti-RAPGEFL1 Mouse Monoclonal Antibody [clone: 4G2]
Vasta-aineet
RAPGEFL1 is a 456 amino acid protein with an estimated molecular weight of ~52 kDa. It is a guanine-nucleotide releasing factor and contains one Ras-GEF domain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RAPGEFL1
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR