Anti-SorCS3 Mouse Monoclonal Antibody [clone: 3B12]

Toimittaja: Biosensis
BSENM-892-100EA 327 EUR
BSENM-892-100
Anti-SorCS3 Mouse Monoclonal Antibody [clone: 3B12]
Vasta-aineet
SorCS3 is a family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. It is noted that all mammalian VPS10 domains carry an N-terminal signal peptide for translocation into the endoplasmic reticulum. The greatest similarity between the receptors is found in the C-terminal parts of the VPS10 domains, which harbor 12 cysteine residues with a characteristic spacing conserved in all VPS10 proteins from yeast to man. SorCS3 is a single-pass type I membrane protein that is highly expressed in brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SorCS3
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR