Anti-Netrin-G2 Mouse Monoclonal Antibody [clone: 4F11]

BSENM-909-100EA 327 EUR
BSENM-909-100
Anti-Netrin-G2 Mouse Monoclonal Antibody [clone: 4F11]
Vasta-aineet
Netrin-G2 promotes neurite outgrowth of both axons and dendrites. It is a cell membrane lipid anchor protein that is N-glycosylated.

Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Netrin-G2
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR