Anti-MBNL1 Protein Mouse Monoclonal Antibody [clone: 1D11]
Toimittaja: Biosensis
BSENM-913-100EA 327 EUR
BSENM-913-100
Anti-MBNL1 Protein Mouse Monoclonal Antibody [clone: 1D11]
Vasta-aineet
MBNL1 protein is a 343 amino acid peptide with an estimated molecular weight of 36.5 kDa. It binds to CUG triplet repeat expansion dsRNA.
Specificity has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MBNL1 Protein
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat