Anti-ATG5 Mouse Monoclonal Antibody [clone: 3D2]

Toimittaja: Biosensis
BSENM-881-100EA 327 EUR
BSENM-881-100
Anti-ATG5 Mouse Monoclonal Antibody [clone: 3D2]
Vasta-aineet
Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. ATG5 conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed. ATG5 may also play an important role in the apoptotic process, possibly within the modified cytoskeleton. Its expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. It localise to the cytoplasm and it co-localise with nonmuscle actin. ATG5 is ubiquitously expressed. Its expression is dramatically increased in apoptotic cells. Conjugation to ATG12 is essential for autophage but it is not required for association with isolation membrane.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ATG5
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR