Anti-Olfactomedin 1 Mouse Monoclonal Antibody [clone: 2G12-1B3]

BSENM-887-100EA 327 EUR
BSENM-887-100
Anti-Olfactomedin 1 Mouse Monoclonal Antibody [clone: 2G12-1B3]
Human olfactomedin 1 (Noelin) shares high sequence homology with the rat protein. While the exact function of the encoded protein is not known, its abundant expression in brain suggests that it may have an essential role in nerve tissue. It localises to the endoplasmic reticulum lumen and there are several isoforms  produced by alternative splicing.

Specificity has been confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Olfactomedin 1
Clonality: Monoclonal
Clone: 2G12-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR