Anti-Fatty acid-binding Protein Chicken Polyclonal Antibody

Toimittaja: Biosensis
BSENC-1528-500EA 345 EUR
BSENC-1528-500
Anti-Fatty acid-binding Protein Chicken Polyclonal Antibody
Vasta-aineet
Fatty acid-binding protein, adipocyte (ALBP) is a lipid transport protein which binds long chain fatty acids and other hydrophobic ligands and delivers them to their receptors in the nucleus. ALBP is found in the cytoplasm and nucleus of adipocytes.

This antibody detects Fatty acid-binding protein, adipocyte in various mouse and rat fat cell lysates at approx 16 kDa.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Fatty acid-binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 71-82 and 97-111 of Human FABP4
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR