Anti-RET Protein Mouse Monoclonal Antibody [clone: 1C6]

Toimittaja: Biosensis
BSENM-918-100EA 327 EUR
BSENM-918-100
Anti-RET Protein Mouse Monoclonal Antibody [clone: 1C6]
Vasta-aineet
RET protein is a member of the cadherin superfamily. It is one of the receptor tyrosine kinases, which are cell-surface molecules that transduce signals for cell growth and differentiation. RET protein plays a crucial role in neural crest development, and it can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in RET protein are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma.

Specificity has been confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RET Protein
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR