Obestatin is generated from the proteolytic cleavage of Ghrelin. Obestatin is a hormone that is produced in specialized epithelial cells of the stomach and small intestine of several mammals including humans.[2] Obestatin was originally identified as an anorectic peptide, but its effect on food intake remains controversial.
The antibody recognizes Obestatin in ELISAs, other applications have not been tested
Application Information:
ELISA . Suggested dilution at 1:500 to 1:2,000 in coating buffer as coating antibody in competitive ELISA testing. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Obestatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Obestatin peptide (76-98 aa) from Mouse ghrelin precursor.
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat