LIM homeobox protein 1 is a potential transcription factor that may play a role in differentiation and development of neural and lymphoid cells. It contains 1 homeobox DNA-binding domain and 2 LIM zinc-binding domains. The latter have a negative regulatory function and disruption of the LIM domains results in an activated form. It localises to the nucleus and is expressed in the brain, thymus and tonsils. It is also expressed in samples from patients with chronic myeloid leukemia and in 58% of acute myeloid leukemia cell lines.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: LIM homeobox Protein 1
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human