BAI2 may be involved in angiogenesis inhibition. It has a similar structure to BAI1 which is a seven-span transmembrane protein. BAI2 is strongly expressed in brain but it is also detected in heart, thymus, skeletal muscle, and diferent cell lines. It is a multi-pass membrane protein. It belongs to the G-protein coupled receptor 2 family and it contains one GPS domain as well as four TSP type-1 domains.
The specificity of this antibody was shown by WB and ELISA against the recombinant partial protein with the GST tag. GST tag alone was used as a negative control.
Application Information:
This antibody is recommended for WB and direct ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BAI2
Clonality: Monoclonal
Clone: 6A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human