The unprocessed precursor of protein AC1 has a length of 93 amino acids and an estimated molecular weight of 10.5KDa. This protein is expressed in neuroblastoma but its function is undetermined.
The specificity of this antibody was shown by ELISA using the recombinant human protein AC1 with GST tag.
Application Information:
This antibody is recommended for ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Protein AC1
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: