Copine 6 belongs to the copine family and contains two C2 domains and one von Willebrand factor A domain. The unprocessed precursor has a length of 557 amino acids and a predicted molecular weight of 62 kDa. This protein exhibits calcium-dependent phospholipid binding properties and is thought to function in membrane trafficking and in synaptic plasticity.
The specificity of this antibody has been confirmed by WB and ELISA against the antigen.
Application Information:
This antibody is recommended for WB, direct and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: copine 6
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: