Anti-Mouse Dicer Protein Chicken Polyclonal Antibody

Toimittaja: Biosensis
BSENC-1557-100EA 312 EUR
BSENC-1557-100
Anti-Mouse Dicer Protein Chicken Polyclonal Antibody
Vasta-aineet
Dicer is the rate limiting enzyme in the formation of mature microRNAs.

Specificity confirmed by IHC and WB in mouse brain.
Application Information:
Western Blotting (WB), Immunohistochemistry (IHC) and Immunoprecipitation (IP). The recommended concentration for IHC in formalin fixed and paraffin embedded tissues and formalin/acetone fixed tissues is 1:200-1:500. For WB, the recommended concentration is 1:1,000 to 1:3,000. Mouse Dicer Protein has a predicted length of 1916 residues and the MW of the monomer is 218 kDa. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Mouse Dicer Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse Dicer protein (1388-1405 aa)
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR