Anti-alpha MSH Guinea Pig Polyclonal Antibody
Toimittaja: Biosensis
BSENGP-030-50EA 312 EUR
BSENGP-030-50
Anti-alpha MSH Guinea Pig Polyclonal Antibody
Vasta-aineet
Proopiomelanocortin (POMC) is the precursor of the melanocortin peptides alpha, beta and gamma. POMC neurons in the arcuate nucleus of the hypothalamus function as key central regulators of food intake and homeostatic control of energy balance in both rodents and non-human primates. MSH increases the pigmentation of skin by increasing the melanin production in melanocytes. MSH is produced in the pituitary gland and belongs to the POMC family.
Specificity was demonstrated by immunohistochemistry.
Application Information:
IHC. A concentration of 1 in 2,000 is recommended for IHC. IHC performed in sheep brain (hypothalamus) demonstrates intense staining of cells. No staining is evident when the primary antibody is pre-absorbed with 0.5mg/ml of alpha MSH. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: alpha MSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 138-150 from Human Proopiomelanocortin (POMC).
Host: Guinea Pig
Isotype: IgG
Reactivity: Human, Rat, Sheep