Anti-TAR DNA-binding Protein 43 Mouse Monoclonal Antibody [clone: 3H8]

Toimittaja: Biosensis
BSENM-1403-100EA 302 EUR
BSENM-1403-100
Anti-TAR DNA-binding Protein 43 Mouse Monoclonal Antibody [clone: 3H8]
Vasta-aineet
TAR DNA-binding protein 43 (TDP43) is a DNA and RNA-binding protein which regulates transcription and splicing (ref: SWISSPROT).

The specificity of this antibody has been confirmed by WB. This antibody detects ~43 kDa TDP43 protein on crude extract of mouse brain nuclear fraction.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:1,000 - 1:5,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TAR DNA-binding Protein 43
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rodent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR