Anti-LPP3 Mouse Monoclonal Antibody [clone: 7H7D3]

Toimittaja: Biosensis
BSENM-1560-100EA 312 EUR
BSENM-1560-100
Anti-LPP3 Mouse Monoclonal Antibody [clone: 7H7D3]
Vasta-aineet
Lipid phosphate phosphohydrolase 3 (LPP3) is a member of the phosphatidic acid phosphatase (PAP) family. LPP3 catalyzes the conversion of phosphatidic acid to diacylglycerol. In addition it hydrolyzes lysophosphatidic acid, ceramide-1-phosphate and sphingosine-1-phosphate (Ref: SWISSPROT).

Confirmed by over-expression of human LPP3 cDNA.
Application Information:
Western Blotting (WB) and Flow cytometry (FACS). For WB, the recommended concentration is 2-3 µg/ml. The LPP3 protein has a predicted length of 311 residues and the MW of the monomer is 35 kDa. For FACS, the recommended concentration is 2.0 µg/ml. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LPP3
Clonality: Monoclonal
Clone: 7H7D3
Conjugation: Unconjugated
Epitope: Human LPP3 (179-196 aa)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR