Anti-Clathrin Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-165-100EA 312 EUR
BSENR-165-100
Anti-Clathrin Rabbit Polyclonal Antibody
Vasta-aineet
Clathrin is a major protein component of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice to either the plasma membrane or the trans-Golgi network. These specialised organelles are involved in the intracellular trafficking of receptors and endocytosis of a variety of macromolecules. Clathrin triskelions, composed of 3 heavy chains and 3 light chains, are the basic subunits of the clathrin coat. In the presence of light chains, hub assembly is influenced by both the pH and the concentration of calcium. Clathrin localises to the cytoplasmic face of coated pit and vesicle membranes.

This antibody has been shown to be specific for clathrin.
Application Information:
IHC, WB. This antibody works in immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 100 to 1: 4000 depending on tissue and detection method. It is usual to use approximately a 10-fold increase for confocal microscopy i.e. a dilution range of 1: 100 to 1: 400.For western blotting a dilution range of 1: 1000 to 1: 8000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Clathrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acids 619-638 of rat clathrin
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR