Peroxiredoxin-2 has a role in redox regulation of the cell.
Antibody recognizes Peroxiredoxin-2 in multiple applications including IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 4,000 is recommended. A dilution of 1: 1,000 to 1: 4,000 is recommended for ELISA. This antibody stains basal cells in rat airways and specific kidney tubule cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity seen in rodent and human, other species not yet tested.
Application Information:
IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 4,000 is recommended. A dilution of 1: 1,000 to 1: 4,000 is recommended for ELISA. This antibody stains basal cells in rat airways and specific kidney tubule cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Peroxiredoxin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: (20-31 aa) from Human Peroxiredoxin-2
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,