FUNCTION: Involved in redox regulation of the cell. Can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. May play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury. SUBUNIT: Homotetramer. SUBCELLULAR LOCATION: Cytoplasm. Lysosome. Also found in lung secretory organelles. MISCELLANEOUS: The active site is the redox-active Cys-47 oxidized to Cys-SOH. Cys-SOH may rapidly react with a Cys-SH of the other subunit to form an intermolecular disulfide with a concomitant homodimer formation. The enzyme may be subsequently regenerated by reduction of the disulfide by thioredoxin . MISCELLANEOUS: Irreversibly inactivated by overoxidation of Cys-47 (to Cys-SO(3)H) upon oxidative stress. SIMILARITY: Belongs to the ahpC/TSA family. Rehydrin subfamily.
This antibody has been shown to be specific for Peroxiredoxin-6 protein.
Application Information:
IHC, WB, ELISA. This antibody works superbly in Immunohistochemistry on frozen or paraffin embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for routine immunohistochemistry are 1: 100 to 1: 1000 depending on tissue and detection method. For western blotting a dilution range of 1: 1000 to 1: 4000 is recommended. A dilution of 1: 1000 to 1: 4000 is recommended for ELISA. This antiserum stains the cytoplasm of epithelial cells in the rat and mouse lung and rat and human brain astrocytes. It stains human brain astrocytes in Parkinson's and Alzheimer's disease and the central core of some Lewy bodies in Parkinson's disease and dementia with Lewy bodies. Other tissues have not yet been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Peroxiredoxin-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human, Mouse