Anti-Somatostatin Rabbit Polyclonal Antibody

Toimittaja: Biosensis
BSENR-1502-50EA 312 EUR
BSENR-1502-50
Anti-Somatostatin Rabbit Polyclonal Antibody
Vasta-aineet
Somatostatin is secreted by neuroendocrine neurons in hypothalamus and interacts with somatostatin receptors inhibiting secretion of somatotropin.

Antibody recognizes Somatostatin in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: highly conserved, human, mouse and rat tested to date, other species are expected because of sequence similarity.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Somatostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Somatostatin peptide (103-116)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR