Anti-GAPD Rabbit Polyclonal Antibody

Toimittaja: AbFrontier
ABFRLF-PA0018AEA 531 EUR
ABFRLF-PA0018A
Anti-GAPD Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GAPD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR