Anti-CREB-A Rabbit Polyclonal Antibody

Toimittaja: AbFrontier
ABFRLF-PA0225EA 267 EUR
ABFRLF-PA0225
Anti-CREB-A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CREB-A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR