ESD-turvakengät, nauhaton, Pure, Elten
Toimittaja: Elten
Pure
114-6238EA 0 EUR
114-6238 114-6242 114-6245
ESD-turvakengät, nauhaton, Pure, Elten
Kengät ja saappaat
Leather-free ESD slip-on shoes made from a hydrophobised microfibre plus a mono-density PU sole (L10).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: White
Sertifikaatit: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.