Anti-Aquaporin 1 Rabbit Polyclonal Antibody

Toimittaja: Boster
BSBTPB9473EA 550 EUR
BSBTPB9473
Anti-Aquaporin 1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Aquaporin-1(AQP1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Aquaporin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR